Merck
  • Combinatorial synthesis with high throughput discovery of protein-resistant membrane surfaces.
Combinatorial synthesis with high throughput discovery of protein-resistant membrane surfaces.
Biomaterials (2013-05-28)
Minghao Gu, Arturo J Vegas, Daniel G Anderson, Robert S Langer, James E Kilduff, Georges Belfort
ABSTRACT
Using combinatorial methods, we synthesized a series of new vinyl amide monomers and graft-polymerized them to light-sensitive poly(ether sulfone) (PES) porous films for protein resistance. To increase the discovery rate and statistical confidence, we developed high throughput surface modification methods (HTP) that allow synthesis, screening and selection of desirable monomers from a large library in a relatively short time (days). A series of amide monomers were synthesized by amidation of methacryloyl chloride with amines and grafted onto commercial poly(ether sulfone) (PES) membranes using irradiation from atmospheric pressure plasma (APP). The modified PES membrane surfaces were then tested and screened for static protein adhesion using HTP. Hydroxyl amide monomers N-(3-hydroxypropyl)methacrylamide (A3), N-(4-hydroxybutyl)methacrylamide (A4), and N-(4-hydroxybutyl)methacrylamide (A6), ethylene glycol (EG) monomer N-(3-methoxypropyl)methacrylamide (A7), and N-(2-(dimethylamino)ethyl)-N-methylmethacrylamide (A8), and N-(2-(diethylamino)ethyl)-N-methylmethacrylamide (A9) all terminated with tertiary amines and were shown to have protein resistance. The PES membranes modified with these monomers exhibited both low protein adhesion (i.e. membrane plugging or fouling) and high flux. Their performance is comparable with previously identified best performing PEG and zwitterionic monomers, i.e. the so-called gold-standard for protein resistance. Combining a Hansen solubility parameter (HSP) analysis of the amide monomers and the HTP filtration results, we conclude that monomer solubility in water correlates with protein-resistant surfaces, presumably through its effects on surface-water interactions.
Sigma-Aldrich
Methacryloyl chloride, 97%, contains  ppm monomethyl ether hydroquinone as stabilizer