Merck
  • Progress in palladium-based catalytic systems for the sustainable synthesis of annulated heterocycles: a focus on indole backbones.
Progress in palladium-based catalytic systems for the sustainable synthesis of annulated heterocycles: a focus on indole backbones.
Chemical Society reviews (2012-03-27)
Mélanie Platon, Régine Amardeil, Laurent Djakovitch, Jean-Cyrille Hierso
ABSTRACT
A survey highlighting the most recent palladium catalytic systems produced and their performances for progress in direct synthesis of indole backbones by heterocarbocyclization of reactive substrates is provided. The discussion is developed in relation with the principles of sustainable chemistry concerning atom and mass economy. In this respect, the general convergent character of the syntheses is of particular interest (one-pot, domino, cascade or tandem reactions), and the substrates accessibility and reactivity, together with the final waste production, are also important. This critical review clearly indicates that the development of ligand chemistry, mainly phosphines and carbenes, in the last few decades gave a significant impetus to powerful functionalization of indoles at virtually all positions of this ubiquitous backbone (118 references).
Sigma-Aldrich
Palladium, powder, <75 μm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder or granules, 99.99% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Sigma-Aldrich
Palladium, sponge, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, 99.995% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%
Palladium, foil, not light tested, 50x50mm, thickness 0.003mm, 99.95%
Palladium, rod, 25mm, diameter 2.0mm, 99.95%
Palladium, wire reel, 1m, diameter 0.025mm, as drawn, 99.9%
Palladium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.95%
Palladium, tube, 50mm, outside diameter 2.0mm, inside diameter 1.6mm, wall thickness 0.2mm, as drawn, 99.95%
Palladium, wire reel, 50m, diameter 0.125mm, hard, 99.95%
Palladium, foil, not light tested, 50x50mm, thickness 0.00075mm, permanent polyester support, 99.75%
Palladium, wire reel, 2m, diameter 0.25mm, as drawn, 99.99+%
Palladium, wire reel, 0.5m, diameter 0.02mm, as drawn, 99.95%
Palladium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99.95%
Palladium, wire reel, 0.1m, diameter 1.0mm, hard, 99.95%
Palladium, foil, not light tested, 50x50mm, thickness 0.002mm, 99.95%
Palladium, foil, not light tested, 25x25mm, thickness 0.0025mm, 99.95%
Palladium, foil, light tested, 25x25mm, thickness 0.006mm, 99.95%
Palladium, foil, not light tested, 100x100mm, thickness 0.005mm, 99.95%
Palladium, wire reel, 0.5m, diameter 0.25mm, hard, 99.95%
Palladium, wire reel, 2m, diameter 0.02mm, as drawn, 99.95%