Merck
Safety of ceftriaxone sodium at extremes of age.
Expert opinion on drug safety (2008-09-02)
Scott V Monte, William Allan Prescott, Kristin K Johnson, Lori Kuhman, Joseph A Paladino
ABSTRACT
Isolated reports of neonatal and infant deaths associated with ceftriaxone-calcium precipitation in the lungs and kidneys have prompted a recommendation from the US FDA in June 2007 advising that in patients of all ages, calcium-containing solutions should not be administered simultaneously or within 48 h of the last ceftriaxone dose. To provide a comprehensive review of the literature surrounding the safety of ceftriaxone in the neonatal (< or = 28 days) and geriatric populations (> or = 65 years). Multi-database literature search for original research articles, review articles and case reports pertaining to safety of ceftriaxone in the neonatal and geriatric populations. Ceftriaxone should be avoided or significantly minimized in neonates (especially those treated concomitantly with intravenous calcium solutions and those with hyperbilirubinemia), and potentially restricted in the geriatric population treated concomitantly with intravenous calcium.
Sigma-Aldrich
Ceftriaxone disodium salt hemi(heptahydrate), third-generation cephalosporin antibiotic
USP
Ceftriaxone sodium, United States Pharmacopeia (USP) Reference Standard
Ceftriaxone sodium, European Pharmacopoeia (EP) Reference Standard
Supelco
Ceftriaxone Sodium, Pharmaceutical Secondary Standard; Certified Reference Material