Merck
Proteomics and physiology of erythritol-producing strains.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2005-01-18)
Yong-Cheol Park, Do-Yup Lee, Dae-Hee Lee, Hyo-Jin Kim, Yeon-Woo Ryu, Jin-Ho Seo
ABSTRACT
In-depth knowledge bases on physiological properties of microbes are required to design a better microbial system at a gene level and to develop an industrially viable process in an optimized scheme. Proteomic analyses of industrially useful microorganisms are particularly important for achieving such objectives. In this review, industrial application of erythritol in food and pharmaceutical areas and proteomic techniques for erythritol-producing microbes were presented. Proteomic technologies for erythritol-producing strains such as Candida magnoliae contained protein or peptide sample preparation for two-dimensional electrophoresis and mass spectrometry, analysis of proteome with matrix assisted laser desorption-ionization/time-of-flight mass spectrometry, liquid chromatography/electrospray ionization/tandem mass spectrometry and similarity searching algorithms. The proteomic information was applied to predict the carbon metabolism of erythritol-synthesizing microorganisms.
Sigma-Aldrich
meso-Erythritol, ≥99% (GC)
Pricing and availability is not currently available.
Erythritol, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.