Merck
alpha-Lactalbumin: structure and function.
FEBS letters (2000-05-20)
E A Permyakov, L J Berliner
ABSTRACT
Small milk protein alpha-lactalbumin (alpha-LA), a component of lactose synthase, is a simple model Ca(2+) binding protein, which does not belong to the EF-hand proteins, and a classical example of molten globule state. It has a strong Ca(2+) binding site, which binds Mg(2+), Mn(2+), Na(+), and K(+), and several distinct Zn(2+) binding sites. The binding of cations to the Ca(2+) site increases protein stability against action of heat and various denaturing agents, while the binding of Zn(2+) to the Ca(2+)-loaded protein decreases its stability. Functioning of alpha-LA requires its interactions with membranes, proteins, peptides and low molecular weight substrates and products. It was shown that these interactions are modulated by the binding of metal cations. Recently it was found that some folding variants of alpha-LA demonstrate bactericidal activity and some of them cause apoptosis of tumor cells.
Sigma-Aldrich
α-Lactalbumin from human milk, ≥95% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Lactalbumin
Natural Bos d 4 (α-lactalbumin), Cow′s milk, pkg of 250 μg, storage temp.:-20°C