Merck
  • Somatostatin receptor-2 negatively regulates β-adrenergic receptor mediated Ca(2+) dependent signaling pathways in H9c2 cells.
Somatostatin receptor-2 negatively regulates β-adrenergic receptor mediated Ca(2+) dependent signaling pathways in H9c2 cells.
Biochimica et biophysica acta (2014-01-15)
Rishi K Somvanshi, Shenglong Zou, Xiaofan Qiu, Ujendra Kumar
ABSTRACT
In the present study, we report that somatostatin receptor 2 (SSTR2) plays a crucial role in modulation of β1AR and β2AR mediated signaling pathways that are associated with increased intracellular Ca(2+) and cardiac complications. In H9c2 cells, SSTR2 colocalizes with β1AR or β2AR in receptor specific manner. SSTR2 selective agonist inhibits isoproterenol and formoterol stimulated cAMP formation and PKA phosphorylation in concentration dependent manner. In the presence of SSTR2 agonist, the expression of PKCα and PKCβ was comparable to the basal condition, however SSTR2 agonist inhibits isoproterenol or formoterol induced PKCα and PKCβ expression, respectively. Furthermore, the activation of SSTR2 not only inhibits calcineurin expression and its activity, but also blocks NFAT dephosphorylation and its nuclear translocation. SSTR2 selective agonist abrogates isoproterenol mediated increase in cell size and protein content (an index of hypertrophy). Taken together, the results described here provide direct evidence in support of cardiac protective role of SSTR2 via modulation of Ca(2+) associated signaling pathways attributed to cardiac hypertrophy.
Product Description
Isoprenaline sulfate, European Pharmacopoeia (EP) Reference Standard
Isoprenaline hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Isoprenaline hydrochloride