Merck
  • Association between antimicrobial consumption and resistance in Escherichia coli.
Association between antimicrobial consumption and resistance in Escherichia coli.
Antimicrobial agents and chemotherapy (2008-12-24)
Miika Bergman, Solja T Nyberg, Pentti Huovinen, Pirkko Paakkari, Antti J Hakanen
ABSTRACT
During a 9-year study period from 1997 through 2005, the association between antimicrobial resistance rates in Escherichia coli and outpatient antimicrobial consumption was investigated in 20 hospital districts in Finland. A total of 754,293 E. coli isolates, mainly from urine samples, were tested for antimicrobial resistance in 26 clinical microbiology laboratories. The following antimicrobials were studied: ampicillin, amoxicillin-clavulanate, cephalosporins, fluoroquinolones, trimethoprim, trimethoprim-sulfamethoxazole, pivmecillinam, and nitrofurantoin. We applied a protocol used in earlier studies in which the level of antimicrobial consumption over 1 year was compared with the level of resistance in the next year. Statistically significant associations were found for nitrofurantoin use versus nitrofurantoin resistance (P < 0.0001), cephalosporin use versus nitrofurantoin resistance (P = 0.0293), amoxicillin use versus fluoroquinolone resistance (P = 0.0031), and fluoroquinolone use versus ampicillin resistance (P = 0.0046). Interestingly, we found only a few associations between resistance and antimicrobial consumption. The majority of the associations studied were not significant, including the association between fluoroquinolone use and fluoroquinolone resistance.
Sigma-Aldrich
Pivmecillinam, ≥98% (HPLC)