Merck
  • Neurosteroid allopregnanolone attenuates high glucose-induced apoptosis and prevents experimental diabetic neuropathic pain: in vitro and in vivo studies.
Neurosteroid allopregnanolone attenuates high glucose-induced apoptosis and prevents experimental diabetic neuropathic pain: in vitro and in vivo studies.
The Journal of steroid biochemistry and molecular biology (2013-11-02)
Samira Afrazi, Saeed Esmaeili-Mahani, Vahid Sheibani, Mehdi Abbasnejad
ABSTRACT
Hyperglycemia plays a critical role in the development of diabetic neuropathy. Hyperglycemia induces oxidative stress in neurons, resulting in neuronal cell apoptosis and dysfunction. Anti-apoptotic properties of neurosteroids have been demonstrated in numerous cellular models of neurodegenerative studies. Here, the protective effects of neurosteroid allopregnanolone were investigated in in vitro and in vivo models of diabetic neuropathy. The data show that glucose decreased the viability of PC12 cells in a concentration-dependent manner. Allopregnanolone at concentrations of 2.5, 5 and 10μM markedly prevented high glucose-induced toxicity in naïve and NGF-treated (neuron-like) PC12 cells. Furthermore, treatment of diabetic rats with allopregnanolone (5 and 20mg/kg) significantly ameliorated diabetic-induced thermal hyperalgesia. Moreover, this neurosteroid inhibited caspase 3 and decreased Bax/Bcl2 ratio in high glucose-treated cells and spinal cord of diabetic rats. In conclusion, the data revealed that allopregnanolone has protective effects against hyperglycemic-induced cellular damage and prevention of cell apoptosis is involved in its mechanisms. Our findings suggest that allopregnanolone has protective effect against pro-apoptotic challenges such as diabetes and hyperglycemia and propose therapeutic potential of neurosteroids in attenuation of diabetic side effects such as neuropathy.
Sigma-Aldrich
5α-Pregnan-3α-ol-20-one, solid
Sigma-Aldrich
5β-Pregnan-3α-ol-20-one