Merck
  • Multicolor tunability and upconversion enhancement of fluoride nanoparticles by oxygen dopant.
Multicolor tunability and upconversion enhancement of fluoride nanoparticles by oxygen dopant.
Nanoscale (2013-07-28)
Wenbin Niu, Suli Wu, Shufen Zhang, Liap Tat Su, Alfred Iing Yoong Tok
ABSTRACT
The ability to manipulate the upconversion luminescence of lanthanide-ion doped fluoride upconversion nanoparticles (UCNPs) is particularly important and highly desired due to their wide applications in color displays, multiplexing bioassays and multicolor imaging. Here, we developed a strategy for simultaneously tuning color output and enhancing upconversion emission of Yb/Er doped fluoride UCNPs, based on adjusting the oxygen doping level. The synthesis of multicolored multifunctional NaGdF4:Yb,Er UCNPs was used as the model host system to demonstrate this protocol. Ammonium nitrate (NH4NO3) was used as the oxygen source and added into the reaction system at the beginning stage of nucleation and growth process of fluoride UCNPs, which facilitates the formation of enough oxygen atoms and the diffusion of these into the fluoride host matrix. The results revealed that multicolour output and upconversion enhancement mainly resulted from the variation of phonon energy and crystal field symmetry of the host lattice, respectively. This strategy can be further expanded to other fluoride host matrices. As an example of an application, multicolored UCNPs were used as a color converter in light emitting diodes, which can effectively convert near-infrared light into visible light. It is expected that these multicolored UCNPs will be promising for applications in multiplexing biodetection, bioimaging (optical and magnetic resonance imaging) and other optical technologies, and the present method for the control of O(2-) doping may also be used in other functional nanomaterials.
Sigma-Aldrich
Ytterbium, powder, ≥99.9% trace rare earth metals basis
Sigma-Aldrich
Ammonium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium nitrate, BioXtra, ≥99.5%
Sigma-Aldrich
Ammonium nitrate, suitable for plant cell culture
Sigma-Aldrich
Ammonium nitrate, ≥99.0%
Sigma-Aldrich
Ammonium nitrate, ACS reagent, ≥98%