Merck
  • Biosynthesis of unnatural bacteriochlorophyll c derivatives esterified with α,ω-diols in the green sulfur photosynthetic bacterium Chlorobaculum tepidum.
Biosynthesis of unnatural bacteriochlorophyll c derivatives esterified with α,ω-diols in the green sulfur photosynthetic bacterium Chlorobaculum tepidum.
Biochemistry (2011-08-19)
Risato Nishimori, Tadashi Mizoguchi, Hitoshi Tamiaki, Shigenori Kashimura, Yoshitaka Saga
ABSTRACT
Unnatural bacteriochlorophyll (BChl) c derivatives possessing a hydroxy group at the terminus of a hydrocarbon chain at the 17-propionate were biosynthesized in the green sulfur photosynthetic bacterium Chlorobaculum tepidum. Addition of exogenous 1,8-octanediol, 1,12-dodecanediol, and 1,16-hexadecanediol in acetone to liquid cultures resulted in accumulation of BChl c monoesterified with the corresponding diols. The relative ratios of the novel BChl c derivatives esterified with 1,8-, 1,12-, and 1,16-diols to totally producing BChl c were 8.2, 50.2, and 57.6% in the cells grown with additive α,ω-diols at concentrations of 1.5, 0.06, and 0.06 mM, respectively, at the final concentration. The homologue composition of BChl c derivatives esterified with these α,ω-diols was similar to that of original, coexisting BChl c esterified with farnesol (BChl c(F)), suggesting that esterification of α,ω-diols occurred at the last step of the BChl c biosynthetic pathway by BChl c synthase, BchK, in the same manner as in BChl c(F). Chlorosomes, which were isolated from cells grown in the presence of exogenous α,ω-diols, contained a ratio and a composition of BChl c derivatives esterified with the diols similar to those in the whole cells, indicating that these BChl c derivatives were actually present in chlorosomes. Q(y) absorption bands of C. tepidum cells containing the novel BChl c derivatives were shifted to a shorter wavelength, although their bandwidths were analogous to those of cells obtained by normal cultivation. Circular dichroism spectra of cells that had BChl c derivatives esterified with α,ω-diols exhibited S-shaped signals in the Q(y) region, whose polarities were the reverse of those of cells grown in the normal medium and by supplementation with neat acetone as a control experiment. These spectral features of C. tepidum possessing BChl c derivatives esterified with α,ω-diols imply that the novel BChl c derivatives possessing a hydroxy group at the terminus of a hydrocarbon chain affect their self-assembly in chlorosomes.
Sigma-Aldrich
1,16-Hexadecanediol, 97%
Pricing and availability is not currently available.