Merck
  • Impact of lowering confirmatory test cutoff value in pre-enlistment urine cannabinoids screening: about five years' experience in the French Gendarmerie.
Impact of lowering confirmatory test cutoff value in pre-enlistment urine cannabinoids screening: about five years' experience in the French Gendarmerie.
Journal of analytical toxicology (2012-08-31)
Yannick Lecompte, Martine Perrin, Sophie Salle, Olivier Roussel
ABSTRACT
The guidelines for screening of urinary cannabinoids require that all specimens testing positive should be confirmed by gas chromatography-mass spectrometry at a confirmatory test cutoff value of 15 ng/mL of 11-nor-9-carboxy-Δ9-tetrahydrocannabinol (THCCOOH). To assess the impact of lowering the confirmatory test cutoff value on the diagnostic sensitivity and efficiency of a cannabinoid testing program, the results of 986 confirmation analyses of positive screening tests, conducted in the framework of medical fitness examinations prior to enlistment in the French Gendarmerie between January 1, 2005, and December 31, 2009, were retrospectively studied. If the confirmatory test cutoff value of THCCOOH is set at 5 ng/mL instead of 15 ng/mL as recommended by guidelines, the number of confirmed results increases by 25.2%. The positive predictive value of the initial screening test rises from 63.9 to 80.0%. Only one true-positive applicant has appealed. His THCCOOH urinary concentration, which was incompatible with passive cannabis smoke exposure, was confirmed by another laboratory. The use of a confirmatory test cutoff value lower than that recommended significantly increases the diagnostic sensitivity of the screening program for urinary cannabinoids without altering its specificity.
Product Description
Supelco
(−)-11-nor-9-Carboxy-Δ9-THC solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
(−)-11-nor-9-Carboxy-Δ9-THC solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®