Merck
  • A novel method to prepare water-dispersible magnetic nanoparticles and their biomedical applications: magnetic capture probe and specific cellular uptake.
A novel method to prepare water-dispersible magnetic nanoparticles and their biomedical applications: magnetic capture probe and specific cellular uptake.
Journal of biomedical materials research. Part A (2008-01-09)
Changjiang Yu, Jianjun Zhao, Yingzhi Guo, Cailing Lu, Xu Ma, Zhongwei Gu
ABSTRACT
A novel and simple method to form water-dispersed magnetic nanoparticles was successfully developed through glucosaminic acid-surface modification of iron oxide nanoparticles. The resultant glucosaminic acid-modified magnetic nanoparticles (GA-MNPs) had not only good uniformity in spherical shape with diameter of about 10-13 nm, but also possessed excellent water-dispersity and stability. In cell culture experiments, the internalization of GA-MNPs into different kinds of cells was observed over a 5-day period. The results indicated that the internalization of GA-MNPs into mouse macrophage cells and mouse embryonic fibroblast cells was not observed after 40 h of culturing. However, the GA-MNPs were internalized quickly into cancer cells after just 24 h of culturing. TEM images of the GA-MNPs uptake in ECA-109 cells were used to study the internalization mechanisms of GA-MNPs and their distribution in ECA-109 cells. Additionally, a water-dispersed magnetic capture probe was prepared by immobilization of oligonucleotides onto GA-MNPs, and the probe was used for detection and separation of their complementary oligonucleotides sequence.
Sigma-Aldrich
D-Glucosaminic acid, ≥98% (TLC)