Merck
  • Application of cuprous oxide synthesized from copper-containing waste liquid to treat aqueous reactive dye.
Application of cuprous oxide synthesized from copper-containing waste liquid to treat aqueous reactive dye.
Water science and technology : a journal of the International Association on Water Pollution Research (2012-04-18)
Chao-Yin Kuo, Chueh-Ying Pai
ABSTRACT
Cuprous oxide that was prepared from Cu-containing waste liquid from a printed circuit board manufacturer was used to carry out Fenton-like and photo-Fenton-like oxidation reactions to decolorize a reactive dye (RB19). A microwave hydrothermal method was applied in the synthesis. The highest recovery rate of Cu from wastewater was 87% and was obtained when the synthesis was performed at a power of 200 W for 15 min. An RB19 decolorization efficiency of 99.9% was achieved when the Fenton-like reaction was conducted with 50 mmol/L H(2)O(2) and 0.9 g/L of Cu(2)O. The decolorization of RB19 was more effective when the system was irradiated with visible light than with UV light. Under irradiation, the decolorization rate was the highest when the decolorization was performed in the RB19 solution that contained 50 mmol/L H(2)O(2) and 0.9 g/L CuO(2) in the Fenton-like and the 365 nm-irradiated photo-Fenton-like reactions. In the case of irradiation under 410 nm visible light, the system was operated with lower H(2)O(2) (30 mmol/L) and Cu(2)O (0.5 g/L) to achieve a decolorization rate higher than 365 nm-irradiated photo-Fenton-like reactions.
Sigma-Aldrich
Remazol Brilliant Blue R, anthraquinone dye