Merck
Metal-organic frameworks as hosts for photochromic guest molecules.
Inorganic chemistry (2013-02-16)
D Hermann, H Emerich, R Lepski, D Schaniel, U Ruschewitz
ABSTRACT
Several metal-organic framework compounds (MOF-5, MIL-68(Ga), MIL-68(In), MIL-53(Al)) were loaded with azobenzene (AZB), as confirmed by XRPD measurements and elemental analysis. By IR spectroscopy, it was shown that the light-induced trans/cis isomerization of AZB in these hybrid host-guest compounds is improved compared to that of solid AZB. A population of the excited cis state up to 30% has been obtained for AZB0.66@MIL-68(In). However, no light-induced trans/cis isomerization was observed for AZB0.5@MIL-53(Al). Structural models obtained from high-resolution synchrotron powder diffraction data show that AZB molecules are densely packed within the channels of MIL-53(Al) so that no trans/cis isomerization can occur. A different situation was observed for AZB in the larger channels of MIL-68(Ga). Thus, this investigation shows the influence of the host material on the switching behavior of the embedded AZB molecules.
Sigma-Aldrich
Gallium, 99.99% trace metals basis
Sigma-Aldrich
Gallium, 99.999% trace metals basis
Sigma-Aldrich
Gallium, 99.9995% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 0.45-0.55 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.25 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Indium, beads, diam. 2-5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Indium, powder, 99.99% trace metals basis
Sigma-Aldrich
Indium, rod, diam. 6 mm, ≥99.999% trace metals basis
Sigma-Aldrich
Indium, bars, 99.95%
Sigma-Aldrich
Indium, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 2.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Azobenzene, 98%
Sigma-Aldrich
Indium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.1 mm, ≥99.995% trace metals basis
Sigma-Aldrich
Indium, pieces, 99.99% trace metals basis
Sigma-Aldrich
Indium, beads, diam. 2-5 mm, 99.999% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.995% trace metals basis