Merck
  • Dechlorination of hexachlorobenzene using lead-iron bimetallic particles.
Dechlorination of hexachlorobenzene using lead-iron bimetallic particles.
Chemosphere (2013-01-01)
Xiaoqin Nie, Jianguo Liu, Dongbei Yue, Xianwei Zeng, Yongfeng Nie
ABSTRACT
Synthesized lead-iron (Pb/Fe) bimetallic particles were applied to dechlorinate hexachlorobenzene (HCB) under various conditions (e.g. bimetal amount, initial pH value, reaction temperature, and reaction duration). The results showed that adding Pb onto Fe benefited the dechlorination of HCB and the bimetal with 1.4% Pb content performed best. The degradation rate decreased regularly as the initial pH value of the aqueous increased from 1.9 to 11.1 except for pH 7.0 where the fastest dechlorination rate emerged. The dechlorination could be enhanced by increasing the amount of Pb/Fe or the reaction temperature. The dechlorination ratio of HCB within 15 min increased from 24.3% to 81.3% when Pb/Fe amount increased from 0.1g to 0.8 g. The dechlorination followed pseudo-first-order kinetics, and the dechlorination rate constants were 0.0027, 0.0064, 0.0157, and 0.0321 min(-1) at 25, 50, 70, and 85 °C, respectively, and the activation energy (E(a)) of the dechlorination by Pb/Fe was 37.86 kJ mol(-1).
Product Description
Supelco
Hexachlorobenzene solution, certified reference material, 1000 μg/mL in acetone
SKU
Pack Size
Availability
Price
Quantity
Supelco
Hexachlorobenzene, PESTANAL®, analytical standard