Merck
  • Deterministic order in surface micro-topologies through sequential wrinkling.
Deterministic order in surface micro-topologies through sequential wrinkling.
Advanced materials (Deerfield Beach, Fla.) (2012-08-24)
Jie Yin, Jose Luis Yagüe, Damien Eggenspieler, Karen K Gleason, Mary C Boyce
ABSTRACT
Ordered herringbone patterns with deterministic long and short wavelengths are created using a sequential wrinkling strategy (SWS). Patterns with a prescribed zig-zag turning angle less than 90° are obtained upon sequential wrinkling of non-equi-biaxial prestrain for the first time. SWS provides a new method for measuring thin-film mechanical properties simply through the wrinkling metrology without measurement of film thickness.
Sigma-Aldrich
2-Hydroxyethyl methacrylate, contains ≤250 ppm monomethyl ether hydroquinone as inhibitor, 97%
Sigma-Aldrich
2-Hydroxyethyl methacrylate, ≥99%, contains ≤50 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
2-Hydroxyethyl methacrylate