Merck
  • Copper-catalyzed desymmetric intramolecular Ullmann C-N coupling: an enantioselective preparation of indolines.
Copper-catalyzed desymmetric intramolecular Ullmann C-N coupling: an enantioselective preparation of indolines.
Journal of the American Chemical Society (2012-08-24)
Fengtao Zhou, Jiajia Guo, Jianguang Liu, Ke Ding, Shouyun Yu, Qian Cai
ABSTRACT
The first highly enantioselective copper-catalyzed intramolecular Ullmann C-N coupling reaction has been developed. The asymmetric desymmetrization of 1,3-bis(2-iodoaryl)propan-2-amines catalyzed by CuI/(R)-BINOL-derived ligands led to the enantioselective formation of indolines in high yields and excellent enantiomeric excesses. This method was also applied to the formation of 1,2,3,4-tetrahydroquinolines in high yields and excellent enantioselectivity.
Sigma-Aldrich
Indoline, ReagentPlus®, 99%