Merck
Polyoxometalate-directed assembly of water-soluble AgCl nanocubes.
Chemical communications (Cambridge, England) (2012-01-19)
Alevtina Neyman, Yifeng Wang, Shelly Sharet, Neta Varsano, Bogdan Botar, Paul Kögerler, Louisa Meshi, Ira A Weinstock
ABSTRACT
"Out-of-pocket" association of Ag(+) to the tetradentate defect site of mono-vacant Keggin and Wells-Dawson polyoxometalate (POM) cluster-anions is used to direct the formation of water-soluble AgCl nanocubes.
Sigma-Aldrich
Silver chloride, 99.999% trace metals basis
Sigma-Aldrich
Silver chloride, ReagentPlus®, 99%
Sigma-Aldrich
Silver chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis