Merck
  • Two distinct intracellular Ca2+-release components act in opposite ways in the regulation of the auxin-dependent MIA biosynthesis in Catharanthus roseus cells.
Two distinct intracellular Ca2+-release components act in opposite ways in the regulation of the auxin-dependent MIA biosynthesis in Catharanthus roseus cells.
Journal of experimental botany (2009-02-17)
Pierre Poutrain, Christian Mazars, Martine Thiersault, Marc Rideau, Olivier Pichon
ABSTRACT
Calcium-mediated signalling is ubiquitous in both animals and plants. Changes in cytoplasmic free Ca(2+) concentration couple diverse arrays of stimuli to their specific responses, the specificity of the stimulus being determined by integrated actions between multiple Ca(2+) mobilization pathways. In this work, a pharmacological approach is reported, aimed at deciphering the role of calcium as a second messenger in the transduction pathway leading to the inhibitory effect of 2,4-dichlorophenoxyacetic acid (2,4-D), in regulating monoterpene indole alkaloid (MIA) biosynthesis in Catharanthus roseus cells. It is demonstrated here that auxin-dependent MIA biosynthesis is differentially regulated by two distinct calcium release components from internal stores in C. roseus showing pharmacological profiles similar to those displayed by animal RyR and IP3 channels. MIA biosynthesis is stimulated by caffeine (Ca(2+)-release activator through RyR channels) and by heparin and TMB8 (Ca(2+)-release inhibitors of IP3 channels) whereas MIA biosynthesis is inhibited by mastoparan (Ca(2+)-release activator of IP3 channels) and by ruthenium red and DHBP (Ca(2+)-release inhibitors of RyR channels). Furthermore, calcium, as 2,4-D, acts on MIA biosynthesis by regulating the monoterpene moiety of the MIA biosynthesis pathway since calcium channel modulators preferentially modulate g10h expression, the gene encoding the enzyme of the secoiridoid monoterpene pathway, that is the major target of 2,4-D action. In addition, the simultaneous use of caffeine (an activator of RyR channel in animals) and TMB8 (an inhibitor of the IP3 channel) in 2,4-D treated cells triggers a synergistic effect on MIA accumulation. This finding suggests an opposite and co-ordinated action of multiple Ca(2+)-release pathways in 2,4-D signal transduction, adding a new level of complexity to calcium signalling in plants and questioning the existence of RyR and IP3 channels in plants.
Sigma-Aldrich
8-(Diethylamino)octyl-3,4,5-trimethoxybenzoate hydrochloride, ≥98%