Merck
  • Solid state differentiation of plasma thiols using a centrifugally activated mercaptobenzothiazole disulfide exchange indicator.
Solid state differentiation of plasma thiols using a centrifugally activated mercaptobenzothiazole disulfide exchange indicator.
Chemical communications (Cambridge, England) (2007-02-01)
Samir Chahine, Callum Livingstone, James Davis
ABSTRACT
The solid state interaction of mono and macromolecular thiols at a disulfide heterocycle is shown to provide a versatile pathway for their speciation.
Sigma-Aldrich
2,2′-Dithiobis(benzothiazole), 99%
Sign Into View Organizational & Contract Pricing