Merck
  • Investigations of low-amplitude radio frequency pulses at and away from rotary resonance conditions for I = 5/2 nuclei.
Investigations of low-amplitude radio frequency pulses at and away from rotary resonance conditions for I = 5/2 nuclei.
Solid state nuclear magnetic resonance (2002-12-10)
John W Logan, Jeffry T Urban, Jamie D Walls, Kwang Hun Lim, Alexej Jerschow, Alexander Pines
ABSTRACT
Additional experimental evidence of rotary resonance effects for multiple-quantum coherence conversion in a spin-5/2 system is presented. Two-dimensional plots of the relative efficiency of MQ excitation and conversion are given as a function of radio frequency (rf) amplitude and pulse width. Data are presented for the excitation of five-quantum coherence (5QC), as well as for 5QC to three-quantum coherence (3QC) conversion, 5QC to IQC (the central transition coherence) conversion, and 3QC to IQC conversion. A two-fold increase in the signal-to-noise ratio is achieved by substituting low amplitude rf pulses in place of hard rf pulses for 5QC excitation and 5QC to 3QC conversion in a mixed multiple-quantum magic angle spinning (MAS) (MMQMAS) experiment. The anisotropic line shape for the low-amplitude rf pulse version of the MMQMAS experiment was observed to be distorted from the MAS line shape. The cause and implications of the distortion are discussed.
Sigma-Aldrich
Aluminum acetylacetonate, 98%
Sigma-Aldrich
Aluminum acetylacetonate, purified by sublimation, 99.999% trace metals basis