Merck
  • Heterologous expression of the Bacillus subtilis (natto) alanine dehydrogenase in Escherichia coli and Lactococcus lactis.
Heterologous expression of the Bacillus subtilis (natto) alanine dehydrogenase in Escherichia coli and Lactococcus lactis.
Microbiological research (2009-09-02)
Wei Ye, Guicheng Huo, Junliang Chen, Fei Liu, Jingyuan Yin, Lijie Yang, Xiaolong Ma
ABSTRACT
The major objective of the present study is to change the alanine production of Lactic acid bacteria by expression of Bacillus subtilis (natto) alanine dehydrogenase (AlaDH), the gene that is not present in Lactic acid. B. subtilis AlaDH gene (ald) was cloned into a pGEX6p-1 and expressed in E. coli JM109. Its enzyme activity was 48.3U/mg at 30 degrees C and 45.2U/mg at 42 degrees C. This ald gene was then cloned into a vector pNZ8148 to generate a vector pNZ8148/ald. The same ald gene was placed downstream of the ldh promoter from Streptococcus thermophilus to generate pNZ273/ldhp/ald. The pNZ8148/ald and pNZ273/ldhp/ald were introduced separately in Lactococcus lactis NZ9000. As a result of over-expressed ald, the production of alanine detected by HPLC in L. lactis NZ9000 carrying pNZ273/ldhp/ald reached 52mug/ml, an approximately 26-fold increase compared to the parent strain L. lactis NZ9000, but not in L. lactis NZ9000 carrying pNZ8148/ald. This study would help strain improvement to be used in dairy fermentation for developing healthy yogurts with sweet taste or other fermented dairy foods.
Sigma-Aldrich
L-Alanine Dehydrogenase from Bacillus subtilis, buffered aqueous glycerol solution, ~30 units/mg protein (Lowry)
Sigma-Aldrich
L-Alanine Dehydrogenase from Bacillus subtilis, ammonium sulfate suspension, ≥20 units/mg protein (Lowry)
Sigma-Aldrich
Alanine Dehydrogenase, recombinant, recombinant, expressed in E. coli, ≥15 U/mg