Merck
  • High glucose impairs cognitive function through inducing mitochondrial calcium overload in Treg cells.
High glucose impairs cognitive function through inducing mitochondrial calcium overload in Treg cells.
iScience (2024-01-16)
Ya Hui, Liuyu Kuang, Yuanmei Zhong, Yunyun Tang, Zhiqiang Xu, Tianpeng Zheng
ABSTRACT
High glucose has been proved to impair cognitive function in type 2 diabetes, but the underlying mechanisms remain elusive. Here, we found that high glucose increased transcription factors' SP1 O-GlcNAcylation in regulatory T (Treg) cells. Glycosylated SP1 further enhanced HDAC2 recruitment and histone deacetylation on Na+/Ca2+/Li+ exchanger (NCLX) promoter, which downregulated NCLX expression and led to mitochondrial calcium overload and oxidative damage, thereby promoting Treg cell dysfunction, M1 microglia polarization, and diabetes-associated cognitive impairment. Importantly, GLP-1 receptor agonist alleviated these deleterious effects via GLP-1-receptor-mediated upregulation of OGA and inhibition of SP1 O-GlcNAcylation in Treg cells. Our study highlighted a link between high-glucose-mediated SP1 O-GlcNAcylation and HDAC2/NCLX signaling in control of mitochondrial calcium concentrations in Treg cells. It also revealed a mechanism for linking Treg cell dysfunction and cognitive impairment in type 2 diabetes and provides an insight into the mechanism underlying the neuroprotective effects of GLP-1 receptor agonist.
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit
Sigma-Aldrich
Anti-Histone Deacetylase 6 (HDAC6) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution