Merck
  • Gαs is dispensable for β-arrestin coupling but dictates GRK selectivity and is predominant for gene expression regulation by β2-adrenergic receptor.
Gαs is dispensable for β-arrestin coupling but dictates GRK selectivity and is predominant for gene expression regulation by β2-adrenergic receptor.
The Journal of biological chemistry (2023-09-30)
Valeria Burghi, Justine S Paradis, Adam Officer, Sendi Rafael Adame-Garcia, Xingyu Wu, Edda S F Matthees, Benjamin Barsi-Rhyne, Dana J Ramms, Lauren Clubb, Monica Acosta, Pablo Tamayo, Michel Bouvier, Asuka Inoue, Mark von Zastrow, Carsten Hoffmann, J Silvio Gutkind
ABSTRACT
β-arrestins play a key role in G protein-coupled receptor (GPCR) internalization, trafficking, and signaling. Whether β-arrestins act independently of G protein-mediated signaling has not been fully elucidated. Studies using genome-editing approaches revealed that whereas G proteins are essential for mitogen-activated protein kinase activation by GPCRs., β-arrestins play a more prominent role in signal compartmentalization. However, in the absence of G proteins, GPCRs may not activate β-arrestins, thereby limiting the ability to distinguish G protein from β-arrestin-mediated signaling events. We used β2-adrenergic receptor (β2AR) and its β2AR-C tail mutant expressed in human embryonic kidney 293 cells wildtype or CRISPR-Cas9 gene edited for Gαs, β-arrestin1/2, or GPCR kinases 2/3/5/6 in combination with arrestin conformational sensors to elucidate the interplay between Gαs and β-arrestins in controlling gene expression. We found that Gαs is not required for β2AR and β-arrestin conformational changes, β-arrestin recruitment, and receptor internalization, but that Gαs dictates the GPCR kinase isoforms involved in β-arrestin recruitment. By RNA-Seq analysis, we found that protein kinase A and mitogen-activated protein kinase gene signatures were activated by stimulation of β2AR in wildtype and β-arrestin1/2-KO cells but absent in Gαs-KO cells. These results were validated by re-expressing Gαs in the corresponding KO cells and silencing β-arrestins in wildtype cells. These findings were extended to cellular systems expressing endogenous levels of β2AR. Overall, our results support that Gs is essential for β2AR-promoted protein kinase A and mitogen-activated protein kinase gene expression signatures, whereas β-arrestins initiate signaling events modulating Gαs-driven nuclear transcriptional activity.
Sigma-Aldrich
Anti-Gsα-Subunit, C-Terminal (385-394) Rabbit pAb, liquid, Calbiochem®
Millipore
EZview Red ANTI-FLAG® M2 Affinity Gel, clone M2