Merck
  • A Novel Chromosomal Translocation Identified due to Complex Genetic Instability in iPSC Generated for Choroideremia.
A Novel Chromosomal Translocation Identified due to Complex Genetic Instability in iPSC Generated for Choroideremia.
Cells (2019-09-14)
Nejla Erkilic, Vincent Gatinois, Simona Torriano, Pauline Bouret, Carla Sanjurjo-Soriano, Valerie De Luca, Krishna Damodar, Nicolas Cereso, Jacques Puechberty, Rocio Sanchez-Alcudia, Christian P Hamel, Carmen Ayuso, Isabelle Meunier, Franck Pellestor, Vasiliki Kalatzis
ABSTRACT
Induced pluripotent stem cells (iPSCs) have revolutionized the study of human diseases as they can renew indefinitely, undergo multi-lineage differentiation, and generate disease-specific models. However, the difficulty of working with iPSCs is that they are prone to genetic instability. Furthermore, genetically unstable iPSCs are often discarded, as they can have unforeseen consequences on pathophysiological or therapeutic read-outs. We generated iPSCs from two brothers of a previously unstudied family affected with the inherited retinal dystrophy choroideremia. We detected complex rearrangements involving chromosomes 12, 20 and/or 5 in the generated iPSCs. Suspecting an underlying chromosomal aberration, we performed karyotype analysis of the original fibroblasts, and of blood cells from additional family members. We identified a novel chromosomal translocation t(12;20)(q24.3;q11.2) segregating in this family. We determined that the translocation was balanced and did not impact subsequent retinal differentiation. We show for the first time that an undetected genetic instability in somatic cells can breed further instability upon reprogramming. Therefore, the detection of chromosomal aberrations in iPSCs should not be disregarded, as they may reveal rearrangements segregating in families. Furthermore, as such rearrangements are often associated with reproductive failure or birth defects, this in turn has important consequences for genetic counseling of family members.
Sigma-Aldrich
Anti-REP-1 Antibody, clone 2F1, clone 2F1, from mouse