Merck
  • Overcoming undesirable CYP1A2 inhibition of pyridylnaphthalene-type aldosterone synthase inhibitors: influence of heteroaryl derivatization on potency and selectivity.
Overcoming undesirable CYP1A2 inhibition of pyridylnaphthalene-type aldosterone synthase inhibitors: influence of heteroaryl derivatization on potency and selectivity.
Journal of medicinal chemistry (2008-08-05)
Ralf Heim, Simon Lucas, Cornelia M Grombein, Christina Ries, Katarzyna E Schewe, Matthias Negri, Ursula Müller-Vieira, Barbara Birk, Rolf W Hartmann
ABSTRACT
Recently, we reported on the development of potent and selective inhibitors of aldosterone synthase (CYP11B2) for the treatment of congestive heart failure and myocardial fibrosis. A major drawback of these nonsteroidal compounds was a strong inhibition of the hepatic drug-metabolizing enzyme CYP1A2. In the present study, we examined the influence of substituents in the heterocycle of lead structures with a naphthalene molecular scaffold to overcome this unwanted side effect. With respect to CYP11B2 inhibition, some substituents induced a dramatic increase in inhibitory potency. The methoxyalkyl derivatives 22 and 26 are the most potent CYP11B2 inhibitors up to now (IC50 = 0.2 nM). Most compounds also clearly discriminated between CYP11B2 and CYP11B1, and the CYP1A2 potency significantly decreased in some cases (e.g., isoquinoline derivative 30 displayed only 6% CYP1A2 inhibition at 2 microM concentration). Furthermore, isoquinoline derivative 28 proved to be capable of passing the gastrointestinal tract and reached the general circulation after peroral administration to male Wistar rats.
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Sigma-Aldrich
Sulfaphenazole, ≥98%