Merck
Hydroalkylation of Olefins To Form Quaternary Carbons.
Journal of the American Chemical Society (2019-04-30)
Samantha A Green, Tucker R Huffman, Ruairí O McCourt, Vincent van der Puyl, Ryan A Shenvi
ABSTRACT
Metal-hydride hydrogen atom transfer (MHAT) functionalizes alkenes with predictable branched (Markovnikov) selectivity. The breadth of these transformations has been confined to π-radical traps; no sp3 electrophiles have been reported. Here we describe a Mn/Ni dual catalytic system that hydroalkylates unactivated olefins with unactivated alkyl halides, yielding aliphatic quaternary carbons.
Sigma-Aldrich
trans-(-)-limonene oxide, ≥95%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
trans-(+)-limonene oxide