Merck
  • Targeted inhibition of activated protein C by a non-active-site inhibitory antibody to treat hemophilia.
Targeted inhibition of activated protein C by a non-active-site inhibitory antibody to treat hemophilia.
Nature communications (2020-06-14)
Xiao-Yan Zhao, Andreas Wilmen, Dongli Wang, Xinquan Wang, Maxine Bauzon, Ji-Yun Kim, Lars Linden, Liang Li, Ursula Egner, Tobias Marquardt, Dieter Moosmayer, Jan Tebbe, Julian Marius Glück, Philipp Ellinger, Kirk McLean, Shujun Yuan, Subramanian Yegneswaran, Xiaoqiao Jiang, Vince Evans, Jian-Ming Gu, Doug Schneider, Ying Zhu, Yifan Xu, Cornell Mallari, Ashley Hesslein, Yan Wang, Nicole Schmidt, Katrin Gutberlet, Christine Ruehl-Fehlert, Alexius Freyberger, Terry Hermiston, Chandra Patel, Derek Sim, Laurent O Mosnier, Volker Laux
ABSTRACT
Activated protein C (APC) is a plasma serine protease with antithrombotic and cytoprotective functions. Based on the hypothesis that specific inhibition of APC's anticoagulant but not its cytoprotective activity can be beneficial for hemophilia therapy, 2 types of inhibitory monoclonal antibodies (mAbs) are tested: A type I active-site binding mAb and a type II mAb binding to an exosite on APC (required for anticoagulant activity) as shown by X-ray crystallography. Both mAbs increase thrombin generation and promote plasma clotting. Type I blocks all APC activities, whereas type II preserves APC's cytoprotective function. In normal monkeys, type I causes many adverse effects including animal death. In contrast, type II is well-tolerated in normal monkeys and shows both acute and prophylactic dose-dependent efficacy in hemophilic monkeys. Our data show that the type II mAb can specifically inhibit APC's anticoagulant function without compromising its cytoprotective function and offers superior therapeutic opportunities for hemophilia.
Sigma-Aldrich
Albumin, Bovine Serum, Fraction V, Fatty Acid-Poor, Endotoxin-Free
Pricing and availability is not currently available.