Merck
  • Solid-phase extraction using chloropropyl functionalized sol-gel hybrid sorbent for simultaneous determination of organophosphorus pesticides in selected fruit samples.
Solid-phase extraction using chloropropyl functionalized sol-gel hybrid sorbent for simultaneous determination of organophosphorus pesticides in selected fruit samples.
Journal of separation science (2020-05-10)
Krishna Veni Veloo, Nur Amirah Syahirah Ibrahim
ABSTRACT
A new sol-gel hybrid methyltrimethoxysilane-chloropropyltriethoxysilane was prepared as sorbent for solid-phase extraction. The extraction efficiency of the prepared sol-gel hybrid methyltrimethoxysilane-chloropropyltriethoxysilane was assessed by using three selected organophosphorus pesticides, namely, chlorpyrifos, profenofos, and malathion. Gas chromatography-mass spectrometry was used for detection of organophosphorus pesticides. Several vital parameters were optimized to identify the best extraction conditions. Under the optimum extraction conditions, solid-phase extraction-methyltrimethoxysilane-chloropropyltriethoxysilane method showed good linearity range (0.05-1 μg/mL) with coefficient of determination more than 0.995. The limits of detection obtained were in the range of 0.01-0.07 μg/mL and limits of quantification ranging from 0.03 to 0.21 μg/mL. The limits of detection obtained for the developed method were 2.3-6.5× lower than the limits of detection of commercial octadecyl silica sorbent. Real samples analysis was carried out by applying the developed method on red apple and purple grape samples. The developed method exhibited good recoveries (88.33-120.7%) with low relative standard deviations ranging from 1.6 to 3.3% compared to commercial octadecyl silica sorbent, which showed acceptable recoveries (70.3-100.2%) and relative standard deviations (6.3-8.8%). The solid-phase extraction-methyltrimethoxysilane-chloropropyltriethoxysilane method is presented as an alternative extraction method for determination of organophosphorus pesticides.
Sigma-Aldrich
(3-Chloropropyl)triethoxysilane, 95%