Merck
  • The Expression of CNS-Specific PPARGC1A Transcripts Is Regulated by Hypoxia and a Variable GT Repeat Polymorphism.
The Expression of CNS-Specific PPARGC1A Transcripts Is Regulated by Hypoxia and a Variable GT Repeat Polymorphism.
Molecular neurobiology (2019-09-01)
Selma M Soyal, Petra Bonova, Markus Kwik, Greta Zara, Simon Auer, Cornelia Scharler, Dirk Strunk, Charity Nofziger, Markus Paulmichl, Wolfgang Patsch
ABSTRACT
PPARGC1A encodes a transcriptional co-activator also termed peroxisome proliferator-activated receptor (PPAR) gamma coactivator 1-alpha (PGC-1α) which orchestrates multiple transcriptional programs. We have recently identified CNS-specific transcripts that are initiated far upstream of the reference gene (RG) promoter. The regulation of these isoforms may be relevant, as experimental and genetic studies implicated the PPARGC1A locus in neurodegenerative diseases. We therefore studied cis- and trans-regulatory elements activating the CNS promoter in comparison to the RG promoter in human neuronal cell lines. A naturally occurring variable guanidine thymidine (GT) repeat polymorphism within a microsatellite region in the proximal CNS promoter increases promoter activity in neuronal cell lines. Both the RG and the CNS promoters are activated by ESRRA, and the PGC-1α isoforms co-activate ESRRA on their own promoters suggesting an autoregulatory feedback loop. The proximal CNS, but not the RG, promoter is induced by FOXA2 and co-activated by PGC-1α resulting in robust activation. Furthermore, the CNS, but not the RG, promoter is targeted by the canonical hypoxia response involving HIF1A. Importantly, the transactivation by HIF1A is modulated by the size of the GT polymorphism. Increased expression of CNS-specific transcripts in response to hypoxia was observed in an established rat model, while RG transcripts encoding the full-length reference protein were not increased. These results suggest a role of the CNS region of the PPARGC1A locus in ischemia and warrant further studies in humans as the activity of the CNS promoter as well as its induction by hypoxia is subject to inter-individual variability due to the GT polymorphism.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose, L-glutamine, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
5-Hydroxydecanoic acid sodium salt, ≥97% (HPLC)
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and trace elements, without HEPES and sodium bicarbonate, powder, suitable for cell culture