Merck
  • StAR-related lipid transfer domain 11 (STARD11)-mediated ceramide transport mediates extracellular vesicle biogenesis.
StAR-related lipid transfer domain 11 (STARD11)-mediated ceramide transport mediates extracellular vesicle biogenesis.
The Journal of biological chemistry (2018-08-25)
Masanori Fukushima, Debanjali Dasgupta, Amy S Mauer, Eiji Kakazu, Kazuhiko Nakao, Harmeet Malhi
ABSTRACT
Extracellular vesicles are important carriers of cellular materials and have critical roles in cell-to-cell communication in both health and disease. Ceramides are implicated in extracellular vesicle biogenesis, yet the cellular machinery that mediates the formation of ceramide-enriched extracellular vesicles remains unknown. We demonstrate here that the ceramide transport protein StAR-related lipid transfer domain 11 (STARD11) mediates the release of palmitate-stimulated extracellular vesicles having features consistent with exosomes. Using palmitate as a model of lipotoxic diseases and as a substrate for ceramide biosynthesis in human and murine liver cell lines and primary mouse hepatocytes, we found that STARD11-deficient cells release fewer extracellular vesicles. Moreover, STARD11 reciprocally regulated exosome ceramide enrichment and cellular ceramide depletion. We further observed that in STARD11 knockout cells intracellular ceramide accumulates and that this apparent inability to transfer cellular ceramide into extracellular vesicles reduces cellular viability. Using endogenous markers, we uncovered structural and functional colocalization of the endoplasmic reticulum (ER), STARD11, and multivesicular bodies. This colocalization increased following palmitate treatment, suggesting a functional association that may mediate ceramide trafficking from the ER to the multivesicular body. However, the size and number of multivesicular bodies were comparable in WT and STARD11-knockout cells. In conclusion, we propose a model of how STARD11 mediates ceramide trafficking in palmitate-treated cells and stimulates exosome biogenesis.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-ApoB Antibody, clone 2G11, clone 2G11, from mouse