Merck
  • Oncogenic Pathways and Loss of the Rab11 GTPase Synergize To Alter Metabolism in Drosophila.
Oncogenic Pathways and Loss of the Rab11 GTPase Synergize To Alter Metabolism in Drosophila.
Genetics (2019-06-20)
Yingchao Nie, Shiyan Yu, Qi Li, Niraj K Nirala, Alla Amcheslavsky, Yvonne J K Edwards, Patrick W Shum, Zhong Jiang, Wei Wang, Biliang Zhang, Nan Gao, Y Tony Ip
ABSTRACT
Colorectal cancer is a complex disease driven by well-established mutations such as APC and other yet to be identified pathways. The GTPase Rab11 regulates endosomal protein trafficking, and previously we showed that loss of Rab11 caused intestinal inflammation and hyperplasia in mice and flies. To test the idea that loss of Rab11 may promote cancer progression, we have analyzed archival human patient tissues and observed that 51 out of 70 colon cancer tissues had lower Rab11 protein staining. By using the Drosophila midgut model, we have found that loss of Rab11 can lead to three changes that may relate to cancer progression. First is the disruption of enterocyte polarity based on staining of the FERM domain protein Coracle. Second is an increased proliferation due to an increased expression of the JAK-STAT pathway ligand Upd3. Third is an increased expression of ImpL2, which is an IGFBP7 homolog and can suppress metabolism. Furthermore, loss of Rab11 can act synergistically with the oncoprotein RasV12 to regulate these cancer-related phenotypes.
Sigma-Aldrich
Anti-IGFBP7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Serum Triglyceride Determination Kit, 1 kit sufficient for 250 tests