Merck
  • Inhibition of Notch activity promotes nonmitotic regeneration of hair cells in the adult mouse utricles.
Inhibition of Notch activity promotes nonmitotic regeneration of hair cells in the adult mouse utricles.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2011-10-28)
Vincent Lin, Justin S Golub, Tot Bui Nguyen, Clifford R Hume, Elizabeth C Oesterle, Jennifer S Stone
ABSTRACT
The capacity of adult mammals to regenerate sensory hair cells is not well defined. To explore early steps in this process, we examined reactivation of a transiently expressed developmental gene, Atoh1, in adult mouse utricles after neomycin-induced hair cell death in culture. Using an adenoviral reporter for Atoh1 enhancer, we found that Atoh1 transcription is activated in some hair cell progenitors (supporting cells) 3 d after neomycin treatment. By 18 d after neomycin, the number of cells with Atoh1 transcriptional activity increased significantly, but few cells acquired hair cell features (i.e., accumulated ATOH1 or myosin VIIa protein or developed stereocilia). Treatment with DAPT, an inhibitor of γ-secretase, reduced notch pathway activity, enhanced Atoh1 transcriptional activity, and dramatically increased the number of Atoh1-expressing cells with hair cell features, but only in the striolar/juxtastriolar region. Similar effects were seen with TAPI-1, an inhibitor of another enzyme required for notch activity (TACE). Division of supporting cells was rare in any control or DAPT-treated utricles. This study shows that mature mammals have a natural capacity to initiate vestibular hair cell regeneration and suggests that regional notch activity is a significant inhibitor of direct transdifferentiation of supporting cells into hair cells following damage.
Sigma-Aldrich
TAPI-1, TAPI-1, CAS 171235-71-5, is a structural analog of TAPI-0 with similar in vitro efficacy for the inhibition of MMPs and TACE. Blocks the shedding of several cell surface proteins.
Sigma-Aldrich
TAPI-1 acetate salt, ≥97% (HPLC)