Merck
  • Newly Identified AhR-Active Compounds in the Sediments of an Industrial Area Using Effect-Directed Analysis.
Newly Identified AhR-Active Compounds in the Sediments of an Industrial Area Using Effect-Directed Analysis.
Environmental science & technology (2019-07-23)
Jaeseong Kim, Seongjin Hong, Jihyun Cha, Junghyun Lee, Taewoo Kim, Sunggyu Lee, Hyo-Bang Moon, Kyung-Hoon Shin, Jin Hur, Jung-Suk Lee, John P Giesy, Jong Seong Khim
ABSTRACT
Effect-directed analysis was used to identify previously unidentified aryl hydrocarbon receptor (AhR) agonists in sediments collected from a highly industrialized area of Ulsan Bay, Korea. The specific objectives were to (i) investigate potent fractions of sediment extracts using the H4IIE-luc bioassay, (ii) determine the concentrations of known AhR agonists (polycyclic aromatic hydrocarbons (PAHs) and styrene oligomers (SOs)), (iii) identify previously unreported AhR agonists in fractions by use of GC-QTOFMS, and (iv) evaluate contributions of individual compounds to overall AhR-mediated potencies, found primarily in fractions containing aromatics with log Kow 5-8. Greater concentrations of PAHs and SOs were also found in those fractions. On the basis of GC-QTOFMS and GC-MSD analyses, 16 candidates for AhR agonists were identified in extracts of sediments. Of these, seven compounds, including 1-methylchrysene, benzo[j]fluoranthene, 3-methylchrysene, 5-methylbenz[a]anthracene, 11H-benzo[b]fluorene, benzo[b]naphtho[2,3-d]furan, and benzo[b]naphtho[2,1-d]thiophene, exhibited significant AhR activity. Relative potency values of newly identified AhR agonists were found to be greater than or comparable to that of benzo[a]pyrene (BaP). The potency balance analysis showed that newly identified AhR agonists explained 0.07-16% of bioassay-derived BaP-EQs. These chemicals were widely distributed in industrial sediments; thus, it is of immediate importance to conduct studies on sources and potential effects of those chemicals.
Sigma-Aldrich
2-Bromo-1-ethyl-pyridinium tetrafluoroborate, ≥97.0% (T)