Skip to Content
Merck
vibrant-m

Zonisamide Related Compound C

vibrant-m
Synonym(s):
N′-(Benzisoxazol-3-ylmethylsulfonyl)-N,N-dimethylformimidamide, Zonisamide N,N-dimethylformimidamide
Empirical Formula (Hill Notation):
C11H13N3O3S
CAS Number:
Molecular Weight:
267.30

Products

Brand
Feature
Formula Weight
Manufacturer
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade