
Nifuroxazide

Synonym(s):
Nifuroxazide, 5-Nitro-2-furaldehyde p-hydroxybenzoylhydrazone
Empirical Formula (Hill Notation):
C12H9N3O5
CAS Number:
Molecular Weight:
275.22
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing