Skip to Content
Merck
Ethyl pyruvate ≥97%, FG

Ethyl pyruvate

Ethyl pyruvate ≥97%, FG
Synonym(s):
Ethyl pyruvate
Linear Formula:
CH3COCOOC2H5
CAS Number:
Molecular Weight:
116.12
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing