
Anti-Human Polyvalent Immunoglobulins (α, γ and μ-chain specific)−Alkaline Phosphatase antibody produced in goat

Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1