Skip to Content
Merck
4-(2-Aminoethyl)pyridine 96%

4-(2-Aminoethyl)pyridine

4-(2-Aminoethyl)pyridine 96%
Empirical Formula (Hill Notation):
C7H10N2
CAS Number:
Molecular Weight:
122.17

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing