Merck
GC Analysis of 1-Phenylethanol Enantiomers on Astec® CHIRALDEX B-PM
GC Analysis of 1-Phenylethanol Enantiomers on Astec® CHIRALDEX™ B-PM suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX B-PM Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
standard
Product No.
Description
Pricing
(R)-(+)-1-Phenylethanol
for chiral derivatization, LiChropur, ≥99.0%
1-Phenylethanol-2,2,2-d3
98 atom % D
1-Phenylethanol-1,2-13C2
99 atom % 13C
1-Phenylethanol
98%
1-Phenyl-d5-ethanol
98 atom % D
(S)-(−)-1-Phenylethanol
≥98.5% (sum of enantiomers, GC)
(R)-(+)-1-Phenylethanol
≥98.5% (sum of enantiomers, GC)
1-Phenylethan-1,2,2,2-d4-ol
98 atom % D
1-Phenylethanol-d10
98 atom % D
1-Phenylethanol-1-13C
99 atom % 13C
1-Phenylethanol-2-13C
99 atom % 13C, 99% (CP)
1-Phenyl-1-ethanol
5000 μg/mL in ethanol: isopropanol (95:5), analytical standard
(S)-(−)-1-Phenylethanol
97%
(R)-1-Phenylethanol
97%
CONDITIONS
column
Astec CHIRALDEX B-PM, 30 m x 0.25 mm I.D., 0.12 μm (76023AST)
oven
120 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 24 psi
injection
1 μL, 80:1 split
sample
3 mg/mL in methanol
peak 1: R(+)-1-phenylethanol
peak 2: S(-)-1-phenylethanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC