Merck
Extraction, purification and analysis of histones.
Nature protocols (2007-06-05)
David Shechter, Holger L Dormann, C David Allis, Sandra B Hake
ABSTRACT
Histone proteins are the major protein components of chromatin, the physiologically relevant form of the genome (or epigenome) in all eukaryotic cells. Chromatin is the substrate of many biological processes, such as gene regulation and transcription, replication, mitosis and apoptosis. Since histones are extensively post-translationally modified, the identification of these covalent marks on canonical and variant histones is crucial for the understanding of their biological significance. Many different biochemical techniques have been developed to purify and separate histone proteins. Here, we present standard protocols for acid extraction and salt extraction of histones from chromatin; separation of extracted histones by reversed-phase HPLC; analysis of histones and their specific post-translational modification profiles by acid urea (AU) gel electrophoresis and the additional separation of non-canonical histone variants by triton AU(TAU) and 2D TAU electrophoresis; and immunoblotting of isolated histone proteins with modification-specific antibodies.
Sigma-Aldrich
Histone H4 Protein, human recombinant, 1 mg, Recombinant human histone H4 produced in E. coli, purified by FPLC. For use in Enzyme Assays & Western Blotting.
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
Protamine sulfate salt from herring, Grade III, histone, free(Millon test)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail Set I, Phosphatase Inhibitor Cocktail Set I contains three inhibitors that will inhibit alkaline phosphatases as well as serine/threonine protein phosphatases such as PP1 and PP2A.