Merck
  • Novel impedimetric dopamine biosensor based on boronic acid functional polythiophene modified electrodes.
Novel impedimetric dopamine biosensor based on boronic acid functional polythiophene modified electrodes.
Materials science & engineering. C, Materials for biological applications (2016-12-28)
Muamer Dervisevic, Mehmet Senel, Emre Cevik
ABSTRACT
In this study we report a new, simple and first impedimetric biosensor based on 3-Thienyl boronic acid for dopamine detection. Biosensor electrode preparation is 1min long by simple electro-polymerization of 3-Thienyl boronic acid and copolymer Thiophene P(TBA
Sigma-Aldrich
3-Thienylboronic acid, ≥95.0%
Sigma-Aldrich
Thiophene, ≥99%