Merck
  • Fusion-triggered switching of enzymatic activity on an artificial cell membrane.
Fusion-triggered switching of enzymatic activity on an artificial cell membrane.
Sensors (Basel, Switzerland) (2012-07-11)
Masaru Mukai, Yoshihiro Sasaki, Jun-ichi Kikuchi
ABSTRACT
A nanosensory membrane device was constructed for detecting liposome fusion through changes in an enzymatic activity. Inspired by a biological signal transduction system, the device design involved functionalized liposomal membranes prepared by self-assembly of the following molecular components: a synthetic peptide lipid and a phospholipid as matrix membrane components, a Schiff's base of pyridoxal 5'-phosphate with phosphatidylethanolamine as a thermo-responsive artificial receptor, NADH-dependent L-lactate dehydrogenase as a signal amplifier, and Cu(2+) ion as a signal mediator between the receptor and enzyme. The enzymatic activity of the membrane device was adjustable by changing the matrix lipid composition, reflecting the thermotropic phase transition behavior of the lipid membranes, which in turn controlled receptor binding affinity toward the enzyme-inhibiting mediator species. When an effective fusogen anionic polymer was added to these cationic liposomes, membrane fusion occurred, and the functionalized liposomal membranes responded with changes in enzymatic activity, thus serving as an effective nanosensory device for liposome fusion detection.
Sigma-Aldrich
1,2-Dimyristoyl-sn-glycero-3-phosphoethanolamine, synthetic, ≥99%
Pricing and availability is not currently available.