Merck
  • Synthesis and characterization of novel 2, 2'-bipyrimidine fluorescent derivative for protein binding.
Synthesis and characterization of novel 2, 2'-bipyrimidine fluorescent derivative for protein binding.
Chemistry Central journal (2011-11-10)
Vikas S Padalkar, Vikas S Patil, N Sekar
ABSTRACT
Fluorescent dyes with biocompatible functional group and good fluorescence behavior are used as biosensor for monitoring different biological processes as well as detection of protein assay. All reported fluorophore used as sensors are having high selectivity and sensitivity but till there is more demand to synthesized new fluorophore which have improved fluorescence properties and good biocompatibility. Novel 4, 4'-(1, 1'-(5-(2-methoxyphenoxy)-[2, 2'-bipyrimidine]-4, 6-diyl)bis(1H-pyrazol-3, 1-diyl)) dianiline fluorescent dye was synthesized by multistep synthesis from 2-phenylacetonitrile, 2-chloropyrimidine and 2-methoxyphenol. This dye has absorption at 379 nm with intense single emission at 497 nm having fairly good quantum yield (0.375) and Stokes shift. The intermediates and dye were characterized by FT-IR, 1H NMR, 13C NMR and Mass spectral analysis. The pyrazole bipyrimidine based fluorescent dye possessing two amino groups suitable for binding with protein is reported. Its utility as a biocompatible conjugate was explained by conjugation with bovine serum albumin. The method is based on direct fluorescence detection of fluorophore-labelled protein before and after conjugation. Purified fluorescent conjugate was subsequently analyzed by fluorimetry. The analysis showed that the tested conjugation reaction yielded fluorescent conjugates of the dye through carbodiimide chemistry. In summery synthesized fluorophore pyrazole-bipyrimidine has very good interaction towards protein bovine serum albumin and it acts as good candidate for protein assay.
Sigma-Aldrich
2-Chloropyrimidine, 95%