- Highly regio- and stereoselective synthesis of alpha-(N-alkyl-N-p-toluenesulfonyl)-beta-bromo-ketones via Ni(OAc)2-catalyzed aminobromination of chalcones.
Highly regio- and stereoselective synthesis of alpha-(N-alkyl-N-p-toluenesulfonyl)-beta-bromo-ketones via Ni(OAc)2-catalyzed aminobromination of chalcones.
Chemical biology & drug design (2010-03-25)
Hao Sun, San-Jun Zhi, Jian-Lin Han, Guigen Li, Yi Pan
PMID20331646
ABSTRACT
The combinations of N-methyl-p-toluenesulfonamide/NBS and N-ethyl-p-toluenesulfonamide/NBS were found to be good nitrogen/halogen resources for the aminohalogenation of alpha,beta-unsaturated ketones in the presence of Ni(OAc)(2) as the catalyst for the synthesis of vicinal haloamino ketone derivatives. The introduction of N-alkyl groups to the nitrogen resources resulted in excellent regio- and stereoselectivity for both electron-donating and electron-withdrawing group-attached unsaturated ketone substrates. The structure of the resulting products has been unambiguously confirmed by X-ray crystal structure analysis.