Merck
  • The BRAF(V600E) causes widespread alterations in gene methylation in the genome of melanoma cells.
The BRAF(V600E) causes widespread alterations in gene methylation in the genome of melanoma cells.
Cell cycle (Georgetown, Tex.) (2011-12-23)
Peng Hou, Dingxie Liu, Jianli Dong, Mingzhao Xing
ABSTRACT
Although BRAF(V600E) is well known to play an important role in the tumorigenesis of melanoma, its molecular mechanism, particularly the epigenetic aspect, has been incompletely understood. Here, we investigated the role of BRAF(V600E) signaling in altering gene methylation in the genome of melanoma cells using a methylated CpG island amplification/CpG island microarray system and searched for genes coupled to the BRAF(V600E) signaling through methylation aberrations. The results indicated that a wide range of genes with broad functions were linked to BRAF(V600E) signaling through their hyper- or hypomethylation. Expression of 59 genes hypermethylated upon BRAF knockdown was selectively tested and found to be largely correspondingly underexpressed, suggesting that these genes were naturally hypomethylated, and overexpressed with BRAF(V600E) in melanoma. This BRAF(V600E)-promoted hypomethylation was confirmed on genes selectively examined in primary melanoma tumors. Some of these genes were functionally tested and demonstrated to play a role in melanoma cell proliferation and invasion. As a mechanism of aberrant gene methylation driven by BRAF(V600E), expression of the DNA methyltransferase 1 and histone methyltransferase EZH2 was profoundly affected by BRAF(V600E). We have thus uncovered a previously unrecognized prominent epigenetic mechanism in the tumorigenesis of melanoma driven by BRAF(V600E). Many of the functionally important genes controlled by the BRAF(V600E) signaling through aberrant methylation may prove to be novel therapeutic targets for melanoma.
Sigma-Aldrich
Anti-NR4A2 (AB2) antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-HLX1 antibody produced in rabbit, IgG fraction of antiserum
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-FGD1 antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.