Merck
  • Application of a Raney-cobalt-mediated tandem reductive cyclization protocol to total syntheses of the Aspidosperma alkaloids (±)-limaspermidine and (±)-1-acetylaspidoalbidine.
Application of a Raney-cobalt-mediated tandem reductive cyclization protocol to total syntheses of the Aspidosperma alkaloids (±)-limaspermidine and (±)-1-acetylaspidoalbidine.
Organic letters (2012-10-31)
Shen H Tan, Martin G Banwell, Anthony C Willis, Tristan A Reekie
ABSTRACT
The racemic modification of the Aspidosperma alkaloid limaspermidine (1) has been prepared in ten steps including one involving a Raney-cobalt-mediated tandem reductive cyclization of nitrile 8 to give the tetracyclic system 9b. Compound (±)-1 has been converted over two steps into (±)-1-acetylaspidoalbidine [(±)-13].
Sigma-Aldrich
Carbazole, ≥95% (GC)
Supelco
Carbazole, VETRANAL®, analytical standard