Merck
  • Development of a sensitive enzyme-linked immunosorbent assay for the detection of fumonisin B₁ in maize.
Development of a sensitive enzyme-linked immunosorbent assay for the detection of fumonisin B₁ in maize.
Toxicon : official journal of the International Society on Toxinology (2012-09-11)
Yajie Sheng, Wenxiao Jiang, Sarah De Saeger, Jianzhong Shen, Suxia Zhang, Zhanhui Wang
ABSTRACT
Fumonisin B₁ (FB₁) is a mycotoxin, mainly produced by Fusarium fungi and present in food and feed. It causes harmful effects on human and animal health. Therefore, it is necessary to develop sensitive and reliable screening methods. In this study, a highly sensitive monoclonal antibody (MAb) against FB₁, clone 2D7, was produced, and the 50% inhibition concentration (IC₅₀) of the MAb was 2.2 ng/mL in buffer. The MAb showed high cross-reactivity with fumonisin B₂ (FB₂), and negligible cross-reactivity with other mycotoxins. A sensitive indirect competitive enzyme-linked immunosorbent assay (icELISA) based on this MAb was developed for the determination of FB₁ in maize. In spiked samples (100, 200 and 500 μg/kg), the average recoveries ranged from 78 ± 11 to 107 ± 4%, and the coefficient of variation ranged from 3 to 15%. The limit of detection of the icELISA was 5.4 μg/kg. This method was compared to liquid chromatography tandem mass spectrometry (LC-MS/MS) using naturally contaminated samples, and the correlation coefficient was above 0.82. These results show the reliability of the icELISA method for the determination of FB₁ in maize.
Product Description
Supelco
Fumonisin B1 solution, ~50 μg/mL in acetonitrile: water, analytical standard
Supelco
Fumonisin B1, reference material, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Fumonisin B1 from Fusarium moniliforme, ≥98% (HPLC)