Merck
  • Alpha 1,3-galactosyltransferase deficiency in pigs increases sialyltransferase activities that potentially raise non-gal xenoantigenicity.
Alpha 1,3-galactosyltransferase deficiency in pigs increases sialyltransferase activities that potentially raise non-gal xenoantigenicity.
Journal of biomedicine & biotechnology (2011-12-02)
Jong-Yi Park, Mi-Ryung Park, Deug-Nam Kwon, Min-Hui Kang, Mihye Oh, Jae-Woong Han, Ssang-Goo Cho, Chankyu Park, Dong-Ku Kim, Hyuk Song, Jae-Wook Oh, Jin-Hoi Kim
ABSTRACT
We examined whether deficiency of the GGTA1 gene in pigs altered the expression of several glycosyltransferase genes. Real-time RT-PCR and glycosyltransferase activity showed that 2 sialyltransferases [α2,3-sialyltransferase (α2,3ST) and α2,6-sialyltransferase (α2,6ST)] in the heterozygote GalT KO liver have higher expression levels and activities compared to controls. Enzyme-linked lectin assays indicated that there were also more sialic acid-containing glycoconjugate epitopes in GalT KO livers than in controls. The elevated level of sialic-acid-containing glycoconjugate epitopes was due to the low level of α-Gal in heterozygote GalT KO livers. Furthermore, proteomics analysis showed that heterozygote GalT KO pigs had a higher expression of NAD+-isocitrate dehydrogenase (IDH), which is related to the CMP-N-acetylneuraminic acid hydroxylase (CMAH) enzyme reaction. These findings suggest the deficiency of GGTA1 gene in pigs results in increased production of N-glycolylneuraminic acid (Neu5Gc) due to an increase of α2,6-sialyltransferase and a CMAH cofactor, NAD+-IDH. This indicates that Neu5Gc may be a critical xenoantigen. The deletion of the CMAH gene in the GalT KO background is expected to further prolong xenograft survival.
Sigma-Aldrich
α-2,6-Sialyltransferase from Photobacterium damsela, recombinant, expressed in E. coli BL21, ≥5 units/mg protein
Sigma-Aldrich
N-Glycolylneuraminic acid, ≥95% (HPLC), semisynthetic