Merck
  • Paradoxical Activation of Oncogenic Signaling as a Cancer Treatment Strategy.
Paradoxical Activation of Oncogenic Signaling as a Cancer Treatment Strategy.
Cancer discovery (2024-03-27)
Matheus Henrique Dias, Anoek Friskes, Siying Wang, Joao M Fernandes Neto, Frank van Gemert, Soufiane Mourragui, Chrysa Papagianni, Hendrik J Kuiken, Sara Mainardi, Daniel Alvarez-Villanueva, Cor Lieftink, Ben Morris, Anna Dekker, Emma van Dijk, Lieke H S Wilms, Marcelo S da Silva, Robin A Jansen, Antonio Mulero-Sánchez, Elke Malzer, August Vidal, Cristina Santos, Ramón Salazar, Rosangela A M Wailemann, Thompson E P Torres, Giulia De Conti, Jonne A Raaijmakers, Petur Snaebjornsson, Shengxian Yuan, Wenxin Qin, John S Kovach, Hugo A Armelin, Hein Te Riele, Alexander van Oudenaarden, Haojie Jin, Roderick L Beijersbergen, Alberto Villanueva, Rene H Medema, Rene Bernards
ABSTRACT
Cancer homeostasis depends on a balance between activated oncogenic pathways driving tumorigenesis and engagement of stress response programs that counteract the inherent toxicity of such aberrant signaling. Although inhibition of oncogenic signaling pathways has been explored extensively, there is increasing evidence that overactivation of the same pathways can also disrupt cancer homeostasis and cause lethality. We show here that inhibition of protein phosphatase 2A (PP2A) hyperactivates multiple oncogenic pathways and engages stress responses in colon cancer cells. Genetic and compound screens identify combined inhibition of PP2A and WEE1 as synergistic in multiple cancer models by collapsing DNA replication and triggering premature mitosis followed by cell death. This combination also suppressed the growth of patient-derived tumors in vivo. Remarkably, acquired resistance to this drug combination suppressed the ability of colon cancer cells to form tumors in vivo. Our data suggest that paradoxical activation of oncogenic signaling can result in tumor-suppressive resistance. Significance: A therapy consisting of deliberate hyperactivation of oncogenic signaling combined with perturbation of the stress responses that result from this is very effective in animal models of colon cancer. Resistance to this therapy is associated with loss of oncogenic signaling and reduced oncogenic capacity, indicative of tumor-suppressive drug resistance.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
Anti-phospho Histone H3 (Ser10) Antibody, Alexa Fluor 488 Conjugate, from rabbit, ALEXA FLUOR 488